Gene Transcripts

Gene Symbol
Nme9
Entrez Gene ID
623534
Gene Name
NME/NM23 family member 9
Gene Aliases
Txndc6
Chromosome Location
Chr. 9: 99456243 - 99479899 on Build GRCm38
UniGene ID
Mm.484967
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001165957.1 NP_001159429.1 2 76

View Full Product Details