Gene Transcripts

Gene Symbol
Vmn1r1
Entrez Gene ID
625823
Gene Name
vomeronasal 1 receptor 1
Gene Aliases
EG625823, Gm6628
Chromosome Location
Chr. 1: 182157178 - 182158098 on Build GRCm38
UniGene ID
Mm.479992
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166728.1 NP_001160200.1 1 885

View Full Product Details