Gene Transcripts
- Gene Symbol
- Nlrp1c-ps
- Entrez Gene ID
- 627984
- Gene Name
- NLR family, pyrin domain containing 1C, pseudogene
- Gene Aliases
- Nalp1c, Nlrp1c, OTTMUSG00000006090
- Chromosome Location
- Chr. 11: 71242430 - 71285232 on Build GRCm38
- UniGene ID
- Mm.390378
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC141384.1 |
- |
1, 2 |
30 |
|
DQ117602.1 |
- |
1, 2 |
34 |
|
View Full Product Details