Gene Transcripts

Gene Symbol
Lce1k
Entrez Gene ID
631101
Gene Name
late cornified envelope 1K
Gene Aliases
Gm7055, Sprrl6
Chromosome Location
Chr. 3: 92806291 - 92807891 on Build GRCm38
UniGene ID
Mm.250525
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001254760.1 NP_001241689.1 2 600
GenBank mRNA AV072477.1 - 1 239

View Full Product Details