Gene Transcripts

Gene Symbol
Matr3
Entrez Gene ID
29150
Gene Name
matrin 3
Gene Aliases
P130/MAT3
Chromosome Location
Chr. 18: 28361228 - 28390764 on Build Rnor_6.0
UniGene ID
Rn.29774
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019149.3 NP_062022.2 13 2405
XM_006254534.2 XP_006254596.1 13 2427
XM_006254535.2 XP_006254597.1 13 2378
GenBank mRNA AB205483.1 - 12 3175
M63485.1 - 13 2390

View Full Product Details