Gene Transcripts

Gene Symbol
Hhla1
Entrez Gene ID
654498
Gene Name
HERV-H LTR-associating 1
Gene Aliases
F930104E18Rik
Chromosome Location
Chr. 15: 65922443 - 65976804 on Build GRCm38
UniGene ID
Mm.364550
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145096.1 NP_001138568.1 16 1548
GenBank mRNA AK158329.1 - 16 1550

View Full Product Details