Gene Transcripts

Gene Symbol
Spt1
Entrez Gene ID
29229
Gene Name
salivary protein 1
Gene Aliases
Chromosome Location
Chr. 7: 145325004 - 145338152 on Build Rnor_6.0
UniGene ID
Rn.9933
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019171.1 NP_062044.1 4 314
GenBank mRNA M33976.1 - 4 314

View Full Product Details