Gene Transcripts

Gene Symbol
Frmpd1
Entrez Gene ID
666060
Gene Name
FERM and PDZ domain containing 1
Gene Aliases
BC031840, mKIAA0967
Chromosome Location
Chr. 4: 45184417 - 45285936 on Build GRCm38
UniGene ID
Mm.24783
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081172.2 NP_001074641.1 2 84
GenBank mRNA BB660211.1 - 2 85
BC158060.1 - 2 48 9007360

View Full Product Details