Gene Transcripts

Gene Symbol
Tas2r103
Entrez Gene ID
667992
Gene Name
taste receptor, type 2, member 103
Gene Aliases
EG667992, T2R3, TRB2, Tas2r10, Tas2r3, mGR03, mt2r63
Chromosome Location
Chr. 6: 133036163 - 133037101 on Build GRCm38
UniGene ID
Mm.479865
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053211.1 NP_444441.1 1 426

View Full Product Details