Gene Transcripts

Gene Symbol
Aagab
Entrez Gene ID
66939
Gene Name
alpha- and gamma-adaptin binding protein
Gene Aliases
2310007F21Rik, p34
Chromosome Location
Chr. 9: 63602629 - 63641897 on Build GRCm38
UniGene ID
Mm.34338
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025857.2 NP_080133.1 4 431
GenBank mRNA AK017714.1 - 4 400
AK159897.1 - 4 432
AK182339.1 - 3 186
AK186664.1 - 3 186
AK192186.1 - 3 187
AK209997.1 - 3 223
AK210563.1 - 3 186
AK214885.1 - 3 223
AK215257.1 - 3 223
BC027311.1 - 4 427 3964905

View Full Product Details