Gene Transcripts
- Gene Symbol
- Parp10
- Entrez Gene ID
- 671535
- Gene Name
- poly (ADP-ribose) polymerase family, member 10
- Gene Aliases
- Chromosome Location
- Chr. 15: 76232995 - 76243440 on Build GRCm38
- UniGene ID
- Mm.373750
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001163575.1 |
NP_001157047.1
|
9, 10 |
2496 |
|
NM_001163576.1 |
NP_001157048.1
|
8, 9 |
2600 |
|
GenBank mRNA |
AK028373.1 |
- |
9, 10 |
2444 |
|
AK035309.1 |
- |
8, 9 |
2620 |
|
AK046755.1 |
- |
8, 9 |
2581 |
|
AK146336.1 |
- |
9, 10 |
2496 |
|
AK153205.1 |
- |
8, 9 |
2337 |
|
AK166291.1 |
- |
9, 10 |
2447 |
|
BC023056.1 |
- |
5, 6 |
1333 |
|
BC024074.1 |
- |
9, 10 |
2453 |
|
BC025608.1 |
- |
5, 6 |
1478 |
|
View Full Product Details