Gene Transcripts

Gene Symbol
Lsm1
Entrez Gene ID
67207
Gene Name
LSM1 homolog, U6 small nuclear RNA associated (S. cerevisiae)
Gene Aliases
2810025O06Rik, CASM
Chromosome Location
Chr. 8: 25785288 - 25803975 on Build GRCm38
UniGene ID
Mm.30198
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026032.1 NP_080308.1 4 1139
GenBank mRNA AK041290.1 - 4 1139

View Full Product Details