Gene Transcripts

Gene Symbol
Trappc3l
Entrez Gene ID
692132
Gene Name
trafficking protein particle complex 3 like
Gene Aliases
Bet3l, ENSMUSG00000071340
Chromosome Location
Chr. 10: 34037597 - 34109815 on Build GRCm38
UniGene ID
Mm.383415
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001162937.1 NP_001156409.1 3 329
GenBank mRNA BC099502.1 - 3 329

View Full Product Details