Gene Transcripts

Gene Symbol
Qpct
Entrez Gene ID
70536
Gene Name
glutaminyl-peptide cyclotransferase (glutaminyl cyclase)
Gene Aliases
5730422A13Rik
Chromosome Location
Chr. 17: 79051906 - 79090268 on Build GRCm38
UniGene ID
Mm.293870
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027455.2 NP_081731.1 3 723
GenBank mRNA AK017598.1 - 3 386
AK037197.1 - 3 376
AK045974.1 - 3 724
AK148394.1 - 3 394
BC020023.1 - 2 294 3491756
BC151027.1 - 3 467 9087940
BC151029.1 - 3 467 9087942

View Full Product Details