Gene Transcripts

Gene Symbol
Osbpl7
Entrez Gene ID
71240
Gene Name
oxysterol binding protein-like 7
Gene Aliases
4933437E18Rik
Chromosome Location
Chr. 11: 97050344 - 97068904 on Build GRCm38
UniGene ID
Mm.190450
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081434.1 NP_001074903.1 23 3493
GenBank mRNA AK154677.1 - 23 3531

View Full Product Details