Gene Transcripts

Gene Symbol
Krtap2-4
Entrez Gene ID
71453
Gene Name
keratin associated protein 2-4
Gene Aliases
5530401P19Rik, KAP2.4, KRTAP2.4
Chromosome Location
Chr. 11: 99614017 - 99614846 on Build GRCm38
UniGene ID
Mm.51510
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027800.1 NP_082076.1 1 666
GenBank mRNA AK017442.1 - 1 666

View Full Product Details