Gene Transcripts

Gene Symbol
Sh2d4a
Entrez Gene ID
72281
Gene Name
SH2 domain containing 4A
Gene Aliases
2210402M20Rik, SH2A
Chromosome Location
Chr. 8: 68276528 - 68347704 on Build GRCm38
UniGene ID
Mm.40974
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028182.1 NP_082458.1 9 2702
GenBank mRNA AK008803.1 - 9 2487
BC023784.1 - 9 1552

View Full Product Details