Gene Transcripts

Gene Symbol
Xpot
Entrez Gene ID
73192
Gene Name
exportin, tRNA (nuclear export receptor for tRNAs)
Gene Aliases
1110004L07Rik, 3110065H13Rik, AI452076, C79645, EXPORTIN-T
Chromosome Location
Chr. 10: 121587380 - 121626316 on Build GRCm38
UniGene ID
Mm.25042
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081056.1 NP_001074525.1 7 876
GenBank mRNA AK029113.1 - 2 329

View Full Product Details