Gene Transcripts

Gene Symbol
Jakmip3
Entrez Gene ID
74004
Gene Name
janus kinase and microtubule interacting protein 3
Gene Aliases
6330417G02Rik, AI593226, mKIAA4091
Chromosome Location
Chr. 7: 138940730 - 139031950 on Build GRCm38
UniGene ID
Mm.334298
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028708.2 NP_082984.1 5385
GenBank mRNA AK132869.1 - 2 2036
AK220482.1 - 5365

View Full Product Details