Gene Transcripts

Gene Symbol
H2afv
Entrez Gene ID
77605
Gene Name
H2A histone family, member V
Gene Aliases
C530002L11Rik, H2a.z-2, H2av
Chromosome Location
Chr. 11: 6427226 - 6444443 on Build GRCm38
UniGene ID
Mm.27624
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029938.1 NP_084214.1 3 276
GenBank mRNA AK021267.1 - 3 199
AK155105.1 - 3 279
AK168110.1 - 3 248
AK178770.1 - 1 79
AK187484.1 - 2 123
BC147293.1 - 2 235 9056353
BC147294.1 - 3 261 9056344
BC147741.1 - 3 261 9056690
BC147751.1 - 3 261 9056700
CJ069840.1 - 3 277

View Full Product Details