Gene Transcripts

Gene Symbol
Dbil5
Entrez Gene ID
59116
Gene Name
diazepam binding inhibitor-like 5
Gene Aliases
Elp
Chromosome Location
Chr. 10: 64375926 - 64376938 on Build Rnor_6.0
UniGene ID
Rn.35510
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021596.2 NP_067607.1 2 345
GenBank mRNA AF128531.1 - 2 347

View Full Product Details