Gene Transcripts

Gene Symbol
FOXA3
Entrez Gene ID
3171
Gene Name
forkhead box A3
Gene Aliases
FKHH3, HNF3G, TCF3G
Chromosome Location
Chr. 19: 45864260 - 45873797 on Build GRCh38
UniGene ID
Hs.36137
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004497.2 NP_004488.2 2 853
GenBank mRNA AK223473.1 - 2 782
BC016024.1 - 2 853 3908052
BT006720.1 - 2 656
L12141.1 - 2 650

View Full Product Details