Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022393.2 | NP_071788.2 | 6 | 718 | |
XM_006246803.2 | XP_006246865.1 | 6 | 756 | ||
XM_008767867.1 | XP_008766089.1 | 6 | 756 | ||
XM_008767868.1 | XP_008766090.1 | 6 | 600 | ||
XM_008767869.1 | XP_008766091.1 | 6 | 597 | ||
GenBank mRNA | BC091151.1 | - | 6 | 718 | |
FQ213042.1 | - | 6 | 590 | ||
FQ221204.1 | - | 6 | 607 | ||
FQ228437.1 | - | 6 | 576 | ||
J05495.1 | - | 6 | 578 |