Gene Transcripts
- Gene Symbol
- Sult1b1
- Entrez Gene ID
- 64305
- Gene Name
- sulfotransferase family, cytosolic, 1B, member 1
- Gene Aliases
- ST1B1
- Chromosome Location
- Chr. 14: 22142412 - 22155246 on Build Rnor_6.0
- UniGene ID
- Rn.1183
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022513.2 |
NP_071958.1
|
2, 3 |
372 |
|
XM_008770056.1 |
XP_008768278.1
|
2, 3 |
246 |
|
GenBank mRNA |
D89375.1 |
- |
2, 3 |
323 |
|
FQ209476.1 |
- |
2, 3 |
341 |
|
FQ209486.1 |
- |
2, 3 |
406 |
|
FQ209579.1 |
- |
2, 3 |
427 |
|
FQ210801.1 |
- |
2, 3 |
414 |
|
FQ212608.1 |
- |
2, 3 |
383 |
|
FQ218295.1 |
- |
2, 3 |
385 |
|
U38419.1 |
- |
2, 3 |
375 |
|
View Full Product Details