Gene Transcripts

Gene Symbol
Acads
Entrez Gene ID
64304
Gene Name
acyl-CoA dehydrogenase, C-2 to C-3 short chain
Gene Aliases
Scad
Chromosome Location
Chr. 12: 47254503 - 47263747 on Build Rnor_6.0
UniGene ID
Rn.1167
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022512.2 NP_071957.1 10 1217
GenBank mRNA BC072545.1 - 10 1236
FQ227562.1 - 4 388
J05030.1 - 10 1218

View Full Product Details