Gene Transcripts

Gene Symbol
Gna11
Entrez Gene ID
81662
Gene Name
guanine nucleotide binding protein, alpha 11
Gene Aliases
Chromosome Location
Chr. 7: 11033400 - 11047284 on Build Rnor_6.0
UniGene ID
Rn.21925
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031033.1 NP_112295.1 3 368
GenBank mRNA AF239674.1 - 3 368

View Full Product Details