Gene Transcripts

Gene Symbol
Psmd1
Entrez Gene ID
83806
Gene Name
proteasome (prosome, macropain) 26S subunit, non-ATPase, 1
Gene Aliases
Chromosome Location
Chr. 9: 93080618 - 93155033 on Build Rnor_6.0
UniGene ID
Rn.2891
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031978.1 NP_114184.1 20, 21 2427
GenBank mRNA AJ006340.1 - 20, 21 2427
BC087654.1 - 20, 21 2512
BC105883.1 - 20, 21 2473
BC128734.1 - 20, 21 2472
BC166420.1 - 20, 21 2468
FQ234100.1 - 9, 10 1029

View Full Product Details