Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_053338.2 | NP_445790.2 | 2, 3 | 547 | |
XM_006255055.2 | XP_006255117.1 | 2, 3 | 629 | ||
GenBank mRNA | CK602305.1 | - | 2, 3 | 519 | |
FM054321.1 | - | 2, 3 | 600 | ||
U12187.1 | - | 2, 3 | 519 |