Gene Transcripts

Gene Symbol
Pip4k2b
Entrez Gene ID
89812
Gene Name
phosphatidylinositol-5-phosphate 4-kinase, type II, beta
Gene Aliases
Pip5k2b
Chromosome Location
Chr. 10: 85655223 - 85684138 on Build Rnor_6.0
UniGene ID
Rn.30025
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053550.1 NP_446002.1 2 355
XM_008768131.1 XP_008766353.1 2 634
GenBank mRNA AF033355.1 - 2 355

View Full Product Details