Gene Transcripts
- Gene Symbol
- Prmt3
- Entrez Gene ID
- 89820
- Gene Name
- protein arginine methyltransferase 3
- Gene Aliases
- Hrmt1l3
- Chromosome Location
- Chr. 1: 105113595 - 105200253 on Build Rnor_6.0
- UniGene ID
- Rn.33389
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053557.1 |
NP_446009.1
|
7, 8 |
785 |
|
GenBank mRNA |
AF059530.1 |
- |
7, 8 |
785 |
|
View Full Product Details