Gene Transcripts

Gene Symbol
CCNC
Entrez Gene ID
892
Gene Name
cyclin C
Gene Aliases
CycC
Chromosome Location
Chr. 6: 99542387 - 99568973 on Build GRCh38
UniGene ID
Hs.430646
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013399.1 NP_001013417.1 3, 4 297
NM_005190.3 NP_005181.2 3, 4 519
XM_006715594.1 XP_006715657.1 3, 4 678
XM_011536232.2 XP_011534534.1 3, 4 678
XM_017011436.1 XP_016866925.1 3, 4 678
GenBank mRNA AB125137.1 - 3, 4 306
AK222768.1 - 3, 4 333
AK223247.1 - 3, 4 320
AK291914.1 - 3, 4 328
AK298558.1 - 2, 3 258
BC010135.2 - 3, 4 352 4106819
BC026272.1 - 3, 4 333
BC041123.1 - 3, 4 502 5759974
BC056153.1 - 3, 4 341 6157106
BM788717.1 - 3, 4 297
M74091.1 - 3, 4 320

View Full Product Details