Gene Transcripts

Gene Symbol
Bcl2l10
Entrez Gene ID
114552
Gene Name
BCL2-like 10 (apoptosis facilitator)
Gene Aliases
Chromosome Location
Chr. 8: 82288705 - 82294750 on Build Rnor_6.0
UniGene ID
Rn.67084
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053733.1 NP_446185.1 1 172
GenBank mRNA AY029163.1 - 1 172

View Full Product Details