Gene Transcripts
- Gene Symbol
- Eif4ebp1
- Entrez Gene ID
- 116636
- Gene Name
- eukaryotic translation initiation factor 4E binding protein 1
- Gene Aliases
- PHAS-I
- Chromosome Location
- Chr. 16: 68954860 - 68968248 on Build Rnor_6.0
- UniGene ID
- Rn.11161
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053857.2 |
NP_446309.1
|
2, 3 |
379 |
|
GenBank mRNA |
U05014.1 |
- |
2, 3 |
379 |
|
View Full Product Details