Gene Transcripts
- Gene Symbol
- Pex6
- Entrez Gene ID
- 117265
- Gene Name
- peroxisomal biogenesis factor 6
- Gene Aliases
- Paf-2, Paf2
- Chromosome Location
- Chr. 9: 16568743 - 16580900 on Build Rnor_6.0
- UniGene ID
- Rn.10675
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_057125.1 |
NP_476466.1
|
4 |
1168 |
|
GenBank mRNA |
D63673.1 |
- |
4 |
1168 |
|
View Full Product Details