Gene Transcripts

Gene Symbol
Smu1
Entrez Gene ID
117541
Gene Name
smu-1 suppressor of mec-8 and unc-52 homolog (C. elegans)
Gene Aliases
Bwd, Smu-1
Chromosome Location
Chr. 5: 57042743 - 57061313 on Build Rnor_6.0
UniGene ID
Rn.54978
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057195.1 NP_476543.1 2 129
GenBank mRNA AY029526.1 - 2 129

View Full Product Details