Gene Transcripts

Gene Symbol
Fcer2
Entrez Gene ID
171075
Gene Name
Fc fragment of IgE, low affinity II, receptor for (CD23)
Gene Aliases
CD23, Fcer2a
Chromosome Location
Chr. 12: 2233772 - 2245324 on Build Rnor_6.0
UniGene ID
Rn.10326
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033924.1 NP_001029096.1 9 617
NM_133550.2 NP_598234.2 8 554
XM_008768939.1 XP_008767161.1 8 781
GenBank mRNA AF381978.1 - 8 503
BC078749.1 - 9 617 7124300
X73579.1 - 7 437

View Full Product Details