Gene Transcripts
- Gene Symbol
- EGFL6
- Entrez Gene ID
- 25975
- Gene Name
- EGF like domain multiple 6
- Gene Aliases
- MAEG, W80
- Chromosome Location
- Chr. X: 13569575 - 13633575 on Build GRCh38
- UniGene ID
- Hs.12844
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001167890.1 |
NP_001161362.1
|
2, 3 |
469 |
|
NM_015507.3 |
NP_056322.2
|
2, 3 |
469 |
|
GenBank mRNA |
AF186084.1 |
- |
2, 3 |
327 |
|
AF193055.1 |
- |
2, 3 |
395 |
|
AJ245671.1 |
- |
2, 3 |
422 |
|
AK075214.1 |
- |
2, 3 |
439 |
|
AK300137.1 |
- |
2, 3 |
205 |
|
AK300236.1 |
- |
2, 3 |
439 |
|
AK315018.1 |
- |
2, 3 |
307 |
|
AL117610.1 |
- |
2, 3 |
199 |
|
AY358333.1 |
- |
2, 3 |
316 |
|
BC038587.1 |
- |
2, 3 |
278 |
5767603 |
DA534733.1 |
- |
2, 3 |
440 |
|
DC380958.1 |
- |
2, 3 |
469 |
|
View Full Product Details