Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001191578.1 | NP_001178507.1 | 18 | 3710 | |
XM_006242875.2 | XP_006242937.1 | 21 | 4183 | ||
XM_006242881.2 | XP_006242943.1 | 20 | 4038 | ||
XM_008766141.1 | XP_008764363.1 | 21 | 4084 | ||
GenBank mRNA | X74226.1 | - | 14 | 2095 |