Gene Transcripts

Gene Symbol
Prr21
Entrez Gene ID
245985
Gene Name
proline rich 21
Gene Aliases
Sgp158
Chromosome Location
Chr. 4: 166040503 - 166045308 on Build Rnor_6.0
UniGene ID
Rn.10110
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139184.1 NP_631923.1 3 978
XM_008763356.1 XP_008761578.1 2 1172
GenBank mRNA L08134.1 - 3 978

View Full Product Details