Gene Transcripts

Gene Symbol
Nmt1
Entrez Gene ID
259274
Gene Name
N-myristoyltransferase 1
Gene Aliases
Chromosome Location
Chr. 10: 91126689 - 91160721 on Build Rnor_6.0
UniGene ID
Rn.830
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_148891.2 NP_683689.1 2 589
GenBank mRNA AJ492222.1 - 2 210
BC097277.1 - 2 589 7457194

View Full Product Details