Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001130012.2 | NP_001123484.1 | 3 | 614 | |
NM_001252073.1 | NP_001239002.1 | 2 | 231 | ||
NM_001252075.1 | NP_001239004.1 | 2 | 225 | ||
NM_001252076.1 | NP_001239005.1 | 2 | 231 | ||
NM_004785.5 | NP_004776.3 | 3 | 614 | ||
XM_005255702.4 | XP_005255759.1 | 2 | 1225 | ||
XM_006720981.1 | XP_006721044.1 | 2 | 2091 | ||
XM_017023894.1 | XP_016879383.1 | 2 | 1217 | ||
XM_017023895.1 | XP_016879384.1 | 3 | 469 | ||
GenBank mRNA | AF004900.1 | - | 3 | 492 | |
AF035771.1 | - | 3 | 476 | ||
BC014513.1 | - | 3 | 587 | ||
BC069014.1 | - | 3 | 583 | ||
BC106001.1 | - | 3 | 590 | 5296143 | |
BQ687174.1 | - | 2 | 109 | ||
BX377861.2 | - | 2 | 217 | ||
CB961565.1 | - | 2 | 252 | ||
DB495688.2 | - | 3 | 595 | ||
U82108.1 | - | 3 | 567 | ||
Z50150.1 | - | 3 | 569 |