Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_030833.1 | NP_110460.1 | 1, 2 | 313 | |
GenBank mRNA | AF164040.1 | - | 1, 2 | 313 | |
BC060563.1 | - | 280 | 6918334 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_008760022.1 | XP_008758244.1 | 3, 4 | 345 | |
XM_008760023.1 | XP_008758245.1 | 2, 3 | 297 | ||
XM_008760024.1 | XP_008758246.1 | 3, 4 | 1627 | ||
XM_008760025.1 | XP_008758247.1 | 3, 4 | 329 | ||
XM_219476.7 | XP_219476.5 | 2, 3 | 1830 |