Gene Transcripts

Gene Symbol
Nrgn
Entrez Gene ID
64356
Gene Name
neurogranin
Gene Aliases
RC3
Chromosome Location
Chr. 8: 40015049 - 40023193 on Build Rnor_6.0
UniGene ID
Rn.11236
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024140.2 NP_077054.1 2 389
GenBank mRNA FQ213074.1 - 2 187
L09119.1 - 2 389

View Full Product Details