Gene Transcripts

Gene Symbol
Vsnl1
Entrez Gene ID
24877
Gene Name
visinin-like 1
Gene Aliases
Nvp1, Ratnvp1
Chromosome Location
Chr. 6: 37001360 - 37121969 on Build Rnor_6.0
UniGene ID
Rn.105934
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012686.2 NP_036818.2 2 317
GenBank mRNA BC092197.1 - 2 317
D10666.1 - 2 325

View Full Product Details