Gene Transcripts

Gene Symbol
TAS2R5
Entrez Gene ID
54429
Gene Name
taste 2 receptor member 5
Gene Aliases
T2R5
Chromosome Location
Chr. 7: 141790217 - 141791366 on Build GRCh38
UniGene ID
Hs.675370
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018980.2 NP_061853.1 1 613
GenBank mRNA BC069509.1 - 1 468
BC095522.1 - 1 613 30706589
BC101586.1 - 1 489 8069092
BC101588.1 - 1 489 8069094

View Full Product Details