Gene Transcripts

Gene Symbol
Aqp8
Entrez Gene ID
11833
Gene Name
aquaporin 8
Gene Aliases
AI255744, AQP-8
Chromosome Location
Chr. 7: 123462294 - 123468003 on Build GRCm38
UniGene ID
Mm.273175
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109045.1 NP_001102515.1 6 1005
NM_007474.2 NP_031500.1 6 1008
GenBank mRNA AF018952.1 - 6 990
AK146660.1 - 5 687
BC010982.1 - 6 950 4224850

View Full Product Details