Gene Transcripts

Gene Symbol
Arf5
Entrez Gene ID
11844
Gene Name
ADP-ribosylation factor 5
Gene Aliases
Chromosome Location
Chr. 6: 28423640 - 28426499 on Build GRCm38
UniGene ID
Mm.389075
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007480.1 NP_031506.1 3, 4 315
GenBank mRNA BC132452.1 - 4, 5 303
BC138756.1 - 4, 5 303 8861778
D87902.1 - 3, 4 315

View Full Product Details