Gene Transcripts

Gene Symbol
NLRC4
Entrez Gene ID
58484
Gene Name
NLR family CARD domain containing 4
Gene Aliases
AIFEC, CARD12, CLAN, CLAN1, CLANA, CLANB, CLANC, CLAND, CLR2.1, FCAS4, IPAF
Chromosome Location
Chr. 2: 32224449 - 32265743 on Build GRCh38
UniGene ID
Hs.574741
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199138.1 NP_001186067.1 3 452
NM_001199139.1 NP_001186068.1 3 335
NM_001302504.1 NP_001289433.1 3 466
NM_021209.4 NP_067032.3 3 466
XM_017004619.1 XP_016860108.1 3 452
GenBank mRNA AF376061.1 - 4 680
AK095467.1 - 3 452
AK292673.1 - 3 437
AK314762.1 - 3 437
AY027787.1 - 3 467
AY027788.1 - 3 467
AY027789.1 - 3 467
AY027790.1 - 3 467
AY032589.1 - 2 226
AY035391.1 - 3 335
AY358152.1 - 3 414
BC031555.1 - 3 422 5179909
DN998613.1 - 3 472

View Full Product Details