Gene Transcripts

Gene Symbol
Bmi1
Entrez Gene ID
12151
Gene Name
Bmi1 polycomb ring finger oncogene
Gene Aliases
AW546694, Bmi-1, Pcgf4
Chromosome Location
Chr. 2: 18677018 - 18686629 on Build GRCm38
UniGene ID
Mm.289584
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007552.4 NP_031578.2 7, 8 938
GenBank mRNA BC053708.1 - 7, 8 564
BC056384.1 - 6, 7 468 6854104
DQ337175.1 - 6, 7 467
M64279.1 - 7, 8 938

View Full Product Details