Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007798.3 | NP_031824.1 | 6 | 645 | |
GenBank mRNA | AK002456.1 | - | 6 | 594 | |
AK083393.1 | - | 6 | 645 | ||
AK147192.1 | - | 6 | 592 | ||
AK149884.1 | - | 6 | 594 | ||
AK151790.1 | - | 6 | 592 | ||
AK153047.1 | - | 6 | 591 | ||
AK159992.1 | - | 6 | 630 | ||
AK167361.1 | - | 6 | 626 | ||
AK170959.1 | - | 6 | 618 | ||
AK179071.1 | - | 2 | 127 | ||
AK190750.1 | - | 2 | 130 | ||
AK191341.1 | - | 2 | 108 | ||
AK191739.1 | - | 2 | 131 | ||
AK191997.1 | - | 2 | 99 | ||
AK192159.1 | - | 2 | 127 | ||
AK192561.1 | - | 2 | 127 | ||
AK192818.1 | - | 1 | 80 | ||
AK194693.1 | - | 2 | 93 | ||
AK195254.1 | - | 2 | 92 | ||
AK196522.1 | - | 1 | 42 | ||
AK196531.1 | - | 2 | 96 | ||
AK197104.1 | - | 2 | 127 | ||
AK198001.1 | - | 2 | 127 | ||
AK198619.1 | - | 2 | 128 | ||
AK198709.1 | - | 2 | 127 | ||
AK200297.1 | - | 2 | 97 | ||
AK201305.1 | - | 2 | 127 | ||
AK201756.1 | - | 2 | 127 | ||
AK202640.1 | - | 2 | 127 | ||
AK203701.1 | - | 2 | 127 | ||
AK205609.1 | - | 2 | 126 | ||
AK209352.1 | - | 2 | 128 | ||
AK210127.1 | - | 2 | 127 | ||
AK213062.1 | - | 2 | 127 | ||
AK213791.1 | - | 2 | 127 | ||
BC006656.1 | - | 6 | 636 | 3500700 | |
S69034.1 | - | 6 | 641 |